Anti-VN1R1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13152
89277-726EA 485.45 USD
89277-726
Anti-VN1R1 Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use at a dilution of 1/100 - 1/200. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: VN1R1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR