Anti-loxl3a Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128244
89329-892EA 589.36 USD
89329-892
Anti-loxl3a Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: IHC (Whole mount): 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: loxl3a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR