Anti-GPCR GPR97 (G Protein-coupled Receptor 97) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12603
89277-626EA 481.52 USD
89277-626
Anti-GPCR GPR97 (G Protein-coupled Receptor 97) Rabbit Polyclonal Antibody
Antibodies
IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ADGRG3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR