Anti-FGF acidic (fibroblast growth Factor 1 (acidic)) Rabbit Polyclonal Antibody (Biotin)

Supplier: GeneTex
GTX12474
89286-086EA 610.83 USD
89286-086
Anti-FGF acidic (fibroblast growth Factor 1 (acidic)) Rabbit Polyclonal Antibody (Biotin)
Antibodies
ELISA: Use at a concentration of 0.15 - 0.3 ?g/ml. Sensitivity = 0.2 ng/wellWB: Use at a concentration of 0.1 - 0.2 ?g/ml. Predicted molecular weight: 18.6 kDa. Sensitivity = 1.5-3.0 ng/laneNot tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FGF1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR