Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYTH2 Mouse Monoclonal Antibody [clone: CYT2-21]

Supplier: Genetex

GTX12421
89316-586EA 325.43 USD InStock
89316-586
Anti-CYTH2 Mouse Monoclonal Antibody [clone: CYT2-21]
Antibodies
ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. WB: Use at a concentration of 5 - 10 μg/ml, this concentration is determined using a whole extract of rat brain. Predicted molecular weight: 40 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CYTH2
Clonality: Monoclonal
Clone: CYT2-21
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty