Anti-Raspberry Bushy Dwarf virus Chicken Polyclonal Antibody

Supplier: GeneTex
GTX12243
89286-066EA 392.01 USD
89286-066
Anti-Raspberry Bushy Dwarf virus Chicken Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Raspberry Bushy Dwarf virus
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR