Anti-KLB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX122197
89314-634EA 1169.55 USD
89314-634
Anti-KLB Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KLB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR