Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-plk1 Mouse Monoclonal Antibody [clone: AZ34]

Supplier: Genetex

GTX12211
89328-236EA 358.96 USD InStock
89328-236
Anti-plk1 Mouse Monoclonal Antibody [clone: AZ34]
Antibodies
ELISA: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. WB: Use at a concentration of 1 μg/ml. Predicted molecular weight: ~68 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: plk1
Clonality: Monoclonal
Clone: AZ34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Xenopus laevis

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty