Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-D type cyclin Rat Monoclonal Antibody [clone: 94B]

Supplier: Genetex
GTX12208
89328-232
Anti-D type cyclin Rat Monoclonal Antibody [clone: 94B]
Type: Primary
Antigen: D type cyclin
Clonality: Monoclonal
Clone: 94B
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity:

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty