Anti-PRMT3 Mouse Monoclonal Antibody [clone: PRMT3-367]

Supplier: GeneTex
GTX12190
89316-580EA 635.01 USD
89316-580
Anti-PRMT3 Mouse Monoclonal Antibody [clone: PRMT3-367]
Antibodies
ELISA: Use at an assay dependent dilution. ICC: Use at an assay dependent dilution. WB: Use at a concentration of 1 - 2 ?g/ml. This concentration is recommended using a whole cell extract of the 293T cell line transfected with human PRMT3. Predicted molecular weight: 58 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: PRMT3
Clonality: Monoclonal
Clone: PRMT3-367
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR