Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-H3F3A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12181
89316-578
Anti-H3F3A Rabbit Polyclonal Antibody
IF: Use at a dilution of 1/1000. WB: Use at a dilution of 1/1000. Detects a band of approximately 17 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user. We advise you to centrifuge this product vial before use.

Type: Primary
Antigen: H3F3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Xenopus laevis, Bovine

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty