Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC7 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12174
89336-976
Anti-HDAC7 Rabbit Polyclonal Antibody
IF: Use at a dilution of 1/50. This dilution is recommended using rat normal rat kidney (NRK) cells. IP: Use at an assay dependent dilution. 1.0-1.5 μg of the antibody immunoprecipitates HDAC7 from a RIPA extract (180 μg) of 293T cells expressing recombinant human HDAC7. WB: Use at a dilution of 1/1000. This dilution is recommended using whole extracts of mouse NIH-3T3 cells. Predicted molecular weight: 105 kDa. Additional bands may be detected in various extract preparations. Detection of HDAC7 is specifically inhibited with the immunizing peptide. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HDAC7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty