Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IKBKE Rabbit Polyclonal Antibody

Supplier: Genetex

GTX12143
89286-058EA 358.65 USD InStock
89286-058
Anti-IKBKE Rabbit Polyclonal Antibody
Antibodies
WB: Use at a concentration of 1 - 2 μg/ml. An 80 kDa band is observed. The antibody has tested negative on mouse NIH 3T3 cells. <br/>Based on the sequence similarity, it may cross react with TBK1. <br/>For optimal results, primary antibody incubations should be performed at room temperature. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: IKBKE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty