Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNF-related Apoptosis-inducing Ligand Mouse Monoclonal Antibody [clone: 55B709.3]

Supplier: Genetex
GTX12124
89317-322
Anti-TNF-related Apoptosis-inducing Ligand Mouse Monoclonal Antibody [clone: 55B709.3]
WB: Use at a concentration of 1 - 2 μg/ml. Detects a band of approximately 32 kDa (predicted molecular weight: 33.7 kDa). For optimal results, primary antibody incubations should be performed at room temperature. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TNF-related Apoptosis-inducing Ligand
Clonality: Monoclonal
Clone: 55B709.3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty