Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TFIIB (general transcription Factor IIB) Mouse Monoclonal Antibody [clone: IIB1]

Supplier: Genetex
GTX12094
89286-828
Anti-TFIIB (general transcription Factor IIB) Mouse Monoclonal Antibody [clone: IIB1]
For Western blotting, the suggested use of this reagent is 0.25 - 2.0 µg/ml. For immunofluorescence microscopy, the suggested use of this reagent is 1 -5 µg /ml.

Type: Primary
Antigen: TFIIB (general transcription Factor IIB)
Clonality: Monoclonal
Clone: IIB1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mammalian

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty