Anti-FOXO4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX12075
89337-278EA 627.4 USD
89337-278
Anti-FOXO4 Rabbit Polyclonal Antibody
Antibodies
IF: Use at a dilution of 1/1000. This dilution is determined by using paraformaldehyde fixed AFX transfected COS-7 cells. WB: Use at a dilution of 1/2000. This dilution is determined using human AFX expressed in transfected COS-7 cell extracts. Predicted molecular weight: 50 kDa. Staining of AFX is specifically inhibited by the AFX immunizing peptide. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: FOXO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR