Anti-T Cell Factor 3 + 4 Mouse Monoclonal Antibody [clone: 6F12-3]

Supplier: GeneTex
GTX12065
89316-076EA 629.74 USD
89316-076
Anti-T Cell Factor 3 + 4 Mouse Monoclonal Antibody [clone: 6F12-3]
Antibodies
Type: Primary
Antigen: T Cell Factor 3 + 4
Clonality: Monoclonal
Clone: 6F12-3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR