Anti-LEF1 Mouse Monoclonal Antibody [clone: REMB6]

Supplier: GeneTex
GTX12035
89286-044EA 609.3 USD
89286-044
Anti-LEF1 Mouse Monoclonal Antibody [clone: REMB6]
Antibodies
WB: Use at a concentration of 1 - 5 ?g/ml, using whole extracts of human Jurkat cells or transiently expressed human Cos-1 cells. Predicted MW 55-57 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LEF1
Clonality: Monoclonal
Clone: REMB6
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR