Anti-ZBTB24 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX120013
89330-792EA 727.73 USD
89330-792
Anti-ZBTB24 Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:5000-1:20000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: ZBTB24
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR