Anti-C9orf43 (chromosome 9 open reading frame 43) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX119982
89324-048EA 582.76 USD
89324-048
Anti-C9orf43 (chromosome 9 open reading frame 43) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: C9orf43 (chromosome 9 open reading frame 43)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR