Anti-OCT2 Mouse Monoclonal Antibody [clone: PT2]

Supplier: GeneTex
GTX11992
89286-820EA 610.48 USD
89286-820
Anti-OCT2 Mouse Monoclonal Antibody [clone: PT2]
Antibodies
WB: Use at a dilution of 1/500 - 1/1000. Detects bands of approximately 58.5, 61 and 62 kDa. Uncharacterized bands of lower and higher molecular weight may also be revealed. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: OCT2
Clonality: Monoclonal
Clone: PT2
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR