Anti-NF1A Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11988
89286-040EA 610.3 USD
89286-040
Anti-NF1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NF1A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR