Anti-JUND Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11982
89286-038EA 610.83 USD
89286-038
Anti-JUND Rabbit Polyclonal Antibody
Antibodies
WB: Use at a dilution of 1/500 - 1/1000. Detects a band of approximately 50 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: JUND
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR