Anti-C10ORF93 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX119794
89323-974EA 597.42 USD
89323-974
Anti-C10ORF93 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C10ORF93
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR