Anti-ATPAF2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX119419
89314-048EA 175.99 USD
89314-048
Anti-ATPAF2 Rabbit Polyclonal Antibody
Antibodies
Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ATPAF2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR