Anti-DR6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11933
89286-028EA 618.83 USD
89286-028
Anti-DR6 Rabbit Polyclonal Antibody
Antibodies
WB: Use at a concentration of 2 - 4 ?g/ml. Detects a band of approximately 66 kDa (predicted molecular weight: 79 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: DR6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR