Anti-c-Ets1 (v-ets erythroblastosis virus E26 oncogene homolog 1 (avian)) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11912
89286-812EA 610.48 USD
89286-812
Anti-c-Ets1 (v-ets erythroblastosis virus E26 oncogene homolog 1 (avian)) Rabbit Polyclonal Antibody
Antibodies
WB: Use at a dilution of 1/500 - 1/1000. Detects a band of approximately 54 kDa. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: c-Ets1 (v-ets erythroblastosis virus E26 oncogene homolog 1 (avian))
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR