Anti-CALM Rabbit Polyclonal Antibody [clone: N1N3]

Supplier: GeneTex
GTX119084
89323-630EA 582.76 USD
89323-630
Anti-CALM Rabbit Polyclonal Antibody [clone: N1N3]
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CALM
Clonality: Polyclonal
Clone: N1N3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR