Anti-C5a Mouse Monoclonal Antibody [clone: 2942]

Supplier: GeneTex
GTX11877
89278-794EA 602.26 USD
89278-794
Anti-C5a Mouse Monoclonal Antibody [clone: 2942]
Antibodies
IA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: C5a
Clonality: Monoclonal
Clone: 2942
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR