Anti-ZNF581 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX118693
89313-800EA 176.03 USD
89313-800
Anti-ZNF581 Rabbit Polyclonal Antibody
Antibodies
Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ZNF581
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR