Anti-ZBTB48 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX118671
89334-298EA 1199.03 USD
89334-298
Anti-ZBTB48 Rabbit Polyclonal Antibody
Antibodies
Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: ZBTB48
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR