Anti-EXOSC5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX118473
89323-388EA 582.76 USD
89323-388
Anti-EXOSC5 Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: EXOSC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR