Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-E2F4 Mouse Monoclonal Antibody [clone: WUF3]

Supplier: Genetex

GTX11845
89286-808EA 425.52 USD InStock
89286-808
Anti-E2F4 Mouse Monoclonal Antibody [clone: WUF3]
Antibodies
GSA: Use at an assay dependent dilution. IP (denatured): Use at an assay dependent dilution. WB: Use at a concentration of 0.5 - 1 µ g/ml. This concentration is determined using whole extract of transfected 293T (human embryonal kidney) cells expressing E2F4. Detects a band of approximately 50 kDa (for natural molecule, may vary in a recombinant preparation). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: E2F4
Clonality: Monoclonal
Clone: WUF3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty