Anti-CAR (Coxsackie virus and adenovirusR) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX118382
89335-726EA 582.76 USD
89335-726
Anti-CAR (Coxsackie virus and adenovirusR) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:1000-1:10000, IHC-P: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CAR (Coxsackie virus and adenovirusR)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR