Anti-zeta Crystallin (Crystallin, zeta (quinone reductase)) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX118270
89323-334EA 582.76 USD
89323-334
Anti-zeta Crystallin (Crystallin, zeta (quinone reductase)) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: zeta Crystallin (Crystallin, zeta (quinone reductase))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR