Anti-CMG1 (intraflagellar transport 74 homolog (Chlamydomonas)) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX118199
89323-306EA 582.76 USD
89323-306
Anti-CMG1 (intraflagellar transport 74 homolog (Chlamydomonas)) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CMG1 (intraflagellar transport 74 homolog (Chlamydomonas))
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR