Anti-AKR1A1 Goat Polyclonal Antibody

Supplier: GeneTex
GTX11802
89261-514EA 614.99 USD
89261-514
Anti-AKR1A1 Goat Polyclonal Antibody
Antibodies
Approx 37kDa band observed in Human Liver and Human Placenta lysates. In transfected HEK293 transiently expressing Human AKR1A1 a band of approx. 40kDa is observed. This band is not observed in the non-transfected HEK293. (calculated MW of 36.6kDa according to NP_006057.1 and NP_697021.1). Recommended concentration: 0.1-0.3µg/ml. In paraffin embedded Human Kidney shows Brush Border staining in PCT. Recommended concentration, 2-3µg/ml. Both reported variants (NP_006057.1and NP_697021.1) represent identical protein

Type: Primary
Antigen: AKR1A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Bovine, Dog, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR