Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Dicarbonyl Reductase (dicarbonyl/L-xylulose reductase) Goat Polyclonal Antibody

Supplier: Genetex
GTX11800
89287-880
Anti-Dicarbonyl Reductase (dicarbonyl/L-xylulose reductase) Goat Polyclonal Antibody
Approx 26kDa band observed in human kidney, liver and testes lysates (calculated MW of 25.9kDa according to NP_057370.1). Recommended for use at 0.1-0.3µg/ml. In paraffin embedded human kidney, stains membrane / cytoplasm in epithelial cells of renal glomeruli and tubules. Recommended concentration, 1-2µg/ml.

Type: Primary
Antigen: Dicarbonyl Reductase (dicarbonyl/L-xylulose reductase)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty