Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TIMP2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11797
89336-834
Anti-TIMP2 Rabbit Polyclonal Antibody
WB: Use at a dilution of 1/1000. Predicted molecular weight: 23 kDa. For immunoblotting with chemiluminescent substrates, a starting dilution of 1/5000 is recommended. Higher antibody concentrations may be necessary for non-human samples. Note: TIMP2 is constitutively produced in low (pg/ml) levels in most cell types in tissue culture. The low protein levels produced often require concentration of the cell culture media to visualize the bands by immunoblotting. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: TIMP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty