Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP27 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11788
89316-068
Anti-MMP27 Rabbit Polyclonal Antibody
WB: Use at a dilution of 1/1000. A starting dilution of 1/5000 is recommended for chemiluminescent substrates. By immunoblotting against the reduced protein, the antibody identifies bands at 60 kDa and 58 kDa, as well as bands at 53 kDa and 50 kDa. Note: Higher antibody dilutions may be necessary for non-human samples. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MMP27
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty