Anti-PCSK2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX11787
89317-892EA 494.58 USD
89317-892
Anti-PCSK2 Rabbit Polyclonal Antibody
Antibodies
Recommended Starting Dilutions:Western Blotting: 1/500 - 1/1000Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: PCSK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Monkey, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR