Anti-HP1 gamma (chromobox homolog 3) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX117561
89323-042EA 582.76 USD
89323-042
Anti-HP1 gamma (chromobox homolog 3) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:5000-1:20000, IHC-P: 1:100-1:100, ICC/IF: 1:100-1:1000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HP1 gamma (chromobox homolog 3)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR