Anti-C1orf57 (nucleoside-triphosphatase, cancer-related) Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX116960
89322-874EA 582.56 USD
89322-874
Anti-C1orf57 (nucleoside-triphosphatase, cancer-related) Rabbit Polyclonal Antibody
Antibodies
Suggested starting dilutions are as follows: WB: 1:500-1:3000, ICC/IF: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: C1orf57 (nucleoside-triphosphatase, cancer-related)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR