Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP26 Rabbit Polyclonal Antibody

Supplier: Genetex

GTX11686
89287-878
Anti-MMP26 Rabbit Polyclonal Antibody
WB: Use at a dilution of 1/1000. A starting dilution of 1/5000 is recommended for chemiluminescent substrates. By immunoblotting against the reduced protein, the antibody identifies bands at 30 kDa (zymogen). Activated MMP26 is not detected, since the propeptide domain is released upon activation. MMP26 (matrilysin 2) does not appear to be produced by most normal quiescent cells, but the treatment of many cell types with the phorbol ester TPA or IL1 stimulates production of MMP26. Because of the low protein levels produced (pg/ml) concentration of cell culture media is often required to visualize the bands by immunoblotting. Note: Higher antibody dilutions may be necessary for non-human samples. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MMP26
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty