Anti-SLC26A6 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX116782
89313-266EA 176.03 USD
89313-266
Anti-SLC26A6 Rabbit Polyclonal Antibody
Antibodies
Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: SLC26A6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR