Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mouse Anti-Rat IgG Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Genetex
GTX11674
89307-130
Mouse Anti-Rat IgG Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: MARG2b-3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty