Anti-IgG Mouse Monoclonal Antibody [clone: MARG2b-3]

Supplier: GeneTex
GTX11673
89307-128EA 250.12 USD
89307-128
Anti-IgG Mouse Monoclonal Antibody [clone: MARG2b-3]
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: MARG2b-3
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR