Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP18 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX11627
89316-034
Anti-MMP18 Rabbit Polyclonal Antibody
ELISA: Use at an assay dependent dilution. IHC: Use at an assay dependent dilution. IP: Use at an assay dependent dilution. WB: Use at a dilution of 1/1000. Detects bands of approximately 51 and 53 kDa, plus smaller activation/breakdown products. MMP18 is produced at very low levels in normal, quiescent Xenopus cells and tissues. Collagenase production can be stimulated by mitogens such as TPA or TNF alpha in receptive cell lines. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: MMP18
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Xenopus laevis

Order Now


 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty