Anti-BAK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX116073
89312-998EA 582.76 USD
89312-998
Anti-BAK Rabbit Polyclonal Antibody
Antibodies
Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: BAK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR