Anti-DUSP8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX116071
89312-994EA 582.76 USD
89312-994
Anti-DUSP8 Rabbit Polyclonal Antibody
Antibodies
Antibody reactive against peptide, detects immunogen. Further validation in progress.

Type: Primary
Antigen: DUSP8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Bovine, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR